Product Details

SNP ID
rs1130319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3498284 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTCCTGTTAAAATTCAACAGCTT[G/T]ATTGTGAAAGAATCCAGAGATCTCA
Phenotype
MIM: 613400
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADI1 PubMed Links

Gene Details

Gene
ADI1
Gene Name
acireductone dioxygenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306077.1 4039 UTR 3 NP_001293006.1
NM_018269.3 4039 UTR 3 NP_060739.2

View Full Product Details