Product Details

SNP ID
rs1345479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:63903097 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAGAGAAATTGAAACTATAAAAA[A/G]GGAAACACACAACAGGTCCAAGAAG
Phenotype
MIM: 614633
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS54 PubMed Links

Gene Details

Gene
VPS54
Gene Name
VPS54, GARP complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005739.1 Intron NP_001005739.1
NM_016516.2 Intron NP_057600.2
XM_006712029.3 Intron XP_006712092.1
XM_017004316.1 Intron XP_016859805.1

View Full Product Details