Product Details

SNP ID
rs1952453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:132376085 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCTTTTTCCCAAATTGGAGAGCC[A/G]GTGAGCTGAGGCCCTCCCATTAGTG
Phenotype
MIM: 300413
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MBNL3 PubMed Links

Gene Details

Gene
MBNL3
Gene Name
muscleblind like splicing regulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170701.1 12931 UTR 3 NP_001164172.1
NM_001170702.1 12931 UTR 3 NP_001164173.1
NM_001170703.1 12931 UTR 3 NP_001164174.1
NM_001170704.1 12931 UTR 3 NP_001164175.1
NM_018388.3 12931 UTR 3 NP_060858.2
NM_133486.2 12931 UTR 3 NP_597846.1
XM_005262431.3 12931 UTR 3 XP_005262488.1
XM_005262434.3 12931 UTR 3 XP_005262491.1
XM_005262436.3 12931 UTR 3 XP_005262493.1
XM_005262437.3 12931 UTR 3 XP_005262494.1
XM_005262438.3 12931 UTR 3 XP_005262495.1
XM_005262439.3 12931 UTR 3 XP_005262496.1
XM_005262440.3 12931 UTR 3 XP_005262497.1
XM_005262441.3 12931 UTR 3 XP_005262498.1
XM_005262442.3 12931 UTR 3 XP_005262499.1
XM_011531364.2 12931 UTR 3 XP_011529666.1
XM_017029632.1 12931 UTR 3 XP_016885121.1
XM_017029633.1 12931 UTR 3 XP_016885122.1
XM_017029634.1 12931 UTR 3 XP_016885123.1
XM_017029635.1 12931 UTR 3 XP_016885124.1
XM_017029636.1 12931 UTR 3 XP_016885125.1
XM_017029637.1 12931 UTR 3 XP_016885126.1
XM_017029638.1 12931 UTR 3 XP_016885127.1
XM_017029639.1 12931 Intron XP_016885128.1
Gene
RAP2C-AS1
Gene Name
RAP2C antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details