Product Details
- SNP ID
-
rs1916351
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:167489910 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGACAAATTCTTATTTATCAAACTT[A/T]TATGTGCATAAGAATCACCTGGAGT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WDR49
PubMed Links
Gene Details
- Gene
- WDR49
- Gene Name
- WD repeat domain 49
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178824.3 |
|
Intron |
|
|
NP_849146.1 |
View Full Product Details