Product Details

SNP ID
rs1913164
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:28900436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATAGACAGCATAATGTCTACTTA[A/G]AACCTATTATATGCTTTGTTCTGTG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HMBOX1 PubMed Links

Gene Details

Gene
HMBOX1
Gene Name
homeobox containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135726.2 Intron NP_001129198.1
NM_001324382.1 Intron NP_001311311.1
NM_001324383.1 Intron NP_001311312.1
NM_001324384.1 Intron NP_001311313.1
NM_001324385.1 Intron NP_001311314.1
NM_001324386.1 Intron NP_001311315.1
NM_001324387.1 Intron NP_001311316.1
NM_001324388.1 Intron NP_001311317.1
NM_001324389.1 Intron NP_001311318.1
NM_001324390.1 Intron NP_001311319.1
NM_001324391.1 Intron NP_001311320.1
NM_001324392.1 Intron NP_001311321.1
NM_001324393.1 Intron NP_001311322.1
NM_001324394.1 Intron NP_001311323.1
NM_001324395.1 Intron NP_001311324.1
NM_024567.4 Intron NP_078843.2
XM_005273635.1 Intron XP_005273692.1
XM_005273641.4 Intron XP_005273698.1
XM_006716398.2 Intron XP_006716461.1
XM_011544642.2 Intron XP_011542944.1
XM_017013823.1 Intron XP_016869312.1
XM_017013824.1 Intron XP_016869313.1
XM_017013825.1 Intron XP_016869314.1

View Full Product Details