Product Details

SNP ID
rs2298866
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:10881385 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATATAATAGTGAGAATGTGAATCC[A/C]GCCTTAATATCCAGTGACAGGGATT
Phenotype
MIM: 168730
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRH1 PubMed Links

Gene Details

Gene
PRH1
Gene Name
proline rich protein HaeIII subfamily 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291314.1 Intron NP_001278243.1
NM_001291315.1 Intron NP_001278244.1
Gene
PRH1-PRR4
Gene Name
PRH1-PRR4 readthrough
There are no transcripts associated with this gene.

View Full Product Details