Product Details

SNP ID
rs2296466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:23104162 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATACAGGGTCTCTTGCAACGTGTG[A/G]GCTGCCTCTTGCCAAGAGTGAGTGG
Phenotype
MIM: 613782
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MSRB2 PubMed Links

Gene Details

Gene
MSRB2
Gene Name
methionine sulfoxide reductase B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012228.3 248 Missense Mutation GAG,GGG E46G NP_036360.3
XM_011519426.2 248 Missense Mutation GAG,GGG E46G XP_011517728.1

View Full Product Details