Product Details
- SNP ID
-
rs2300056
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:18157272 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCCTCGGCCTGAAGGAGCAGGAGG[C/T]GGCCCCGTCTGGGCTCCCAGCCAGA
- Phenotype
-
MIM: 612258
MIM: 603157
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MAST3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116015407] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MAST3
- Gene Name
- microtubule associated serine/threonine kinase 3
There are no transcripts associated with this gene.
- Gene
- PIK3R2
- Gene Name
- phosphoinositide-3-kinase regulatory subunit 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005027.3 |
|
Intron |
|
|
NP_005018.1 |
View Full Product Details