Product Details

SNP ID
rs3008862
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:66934143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGAGATTTTTTTAACAGTCCTAG[A/G]TTTAAAAAAATAAATTTAAACATAT
Phenotype
MIM: 616848
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIER1 PubMed Links

Gene Details

Gene
MIER1
Gene Name
MIER1 transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077700.2 Intron NP_001071168.2
NM_001077701.2 Intron NP_001071169.1
NM_001077702.2 Intron NP_001071170.2
NM_001077703.2 Intron NP_001071171.2
NM_001077704.2 Intron NP_001071172.1
NM_001146110.1 Intron NP_001139582.1
NM_001146111.1 Intron NP_001139583.1
NM_001146112.1 Intron NP_001139584.1
NM_001146113.1 Intron NP_001139585.1
NM_001278215.1 Intron NP_001265144.1
NM_020948.3 Intron NP_065999.2
XM_005271076.3 Intron XP_005271133.1
XM_011541864.2 Intron XP_011540166.1
XM_011541865.2 Intron XP_011540167.1
XM_017001919.1 Intron XP_016857408.1
XM_017001920.1 Intron XP_016857409.1
XM_017001921.1 Intron XP_016857410.1
XM_017001922.1 Intron XP_016857411.1
XM_017001923.1 Intron XP_016857412.1
XM_017001924.1 Intron XP_016857413.1
XM_017001925.1 Intron XP_016857414.1
XM_017001926.1 Intron XP_016857415.1
XM_017001927.1 Intron XP_016857416.1
XM_017001928.1 Intron XP_016857417.1
XM_017001929.1 Intron XP_016857418.1
XM_017001930.1 Intron XP_016857419.1
XM_017001931.1 Intron XP_016857420.1
XM_017001932.1 Intron XP_016857421.1
XM_017001933.1 Intron XP_016857422.1
XM_017001934.1 Intron XP_016857423.1
Gene
WDR78
Gene Name
WD repeat domain 78
There are no transcripts associated with this gene.

View Full Product Details