Product Details

SNP ID
rs2997657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:70047773 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAGTTGGGACACAAATCTGCGAA[C/T]TGGGGTTTCATCACAGGAGAATCAA
Phenotype
MIM: 612879
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAMDC2 PubMed Links

Gene Details

Gene
MAMDC2
Gene Name
MAM domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153267.4 Intron NP_694999.3
XM_011518471.1 Intron XP_011516773.1
XM_011518475.2 Intron XP_011516777.1
XM_017014587.1 Intron XP_016870076.1

View Full Product Details