Product Details

SNP ID
rs2301748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31403620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCTTGAGCCGCTGAGAGGGTGG[C/T]GACGTCGGGGCCATGGGGCTGGGCC
Phenotype
MIM: 600169
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MICA PubMed Links

Gene Details

Gene
MICA
Gene Name
MHC class I polypeptide-related sequence A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000247.2 54 UTR 5 NP_000238.1
NM_001177519.2 54 UTR 5 NP_001170990.1
NM_001289152.1 54 Intron NP_001276081.1
NM_001289153.1 54 Intron NP_001276082.1
NM_001289154.1 54 Intron NP_001276083.1

View Full Product Details