Product Details

SNP ID
rs2301805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132883710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAGAGCTACAATTTCTTCTAAA[G/T]AAGTACTATAATCTTGTAATTCATA
Phenotype
MIM: 604417 MIM: 611373
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AFF4 PubMed Links

Gene Details

Gene
AFF4
Gene Name
AF4/FMR2 family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014423.3 Intron NP_055238.1
XM_005271963.4 Intron XP_005272020.1
XM_005271964.4 Intron XP_005272021.1
XM_006714587.3 Intron XP_006714650.1
Gene
LEAP2
Gene Name
liver enriched antimicrobial peptide 2
There are no transcripts associated with this gene.

View Full Product Details