Product Details

SNP ID
rs2301861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103297437 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGACCCCGGAAGCACATCCTTCAT[C/T]CTCTACCTTCCTTCTAGCAGAAATG
Phenotype
MIM: 603131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PMPCB PubMed Links

Gene Details

Gene
PMPCB
Gene Name
peptidase, mitochondrial processing beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004279.2 12 UTR 5 NP_004270.2
XM_005250717.3 12 UTR 5 XP_005250774.1
XM_006716181.3 12 UTR 5 XP_006716244.1

View Full Product Details