Product Details

SNP ID
rs2302053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241190030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACCCATCCCCTCTCTGACCCCC[A/G]TCCCCACCCGGCCTTGCTGGGTGCA
Phenotype
MIM: 605096 MIM: 602877
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANO7 PubMed Links

Gene Details

Gene
ANO7
Gene Name
anoctamin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001666.3 Intron NP_001001666.1
NM_001001891.3 Intron NP_001001891.2
XM_011511263.2 Intron XP_011509565.1
XM_011511267.2 Intron XP_011509569.1
XM_017004229.1 Intron XP_016859718.1
XM_017004230.1 Intron XP_016859719.1
Gene
PPP1R7
Gene Name
protein phosphatase 1 regulatory subunit 7
There are no transcripts associated with this gene.

View Full Product Details