Product Details

SNP ID
rs2302261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64224789 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCCATCTCGTCTCGCACTTGTCC[C/G]CTCACCCCGACCTGCAGGGAATGGC
Phenotype
MIM: 604189 MIM: 607901 MIM: 610470 MIM: 601398
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC4 PubMed Links

Gene Details

Gene
DNAJC4
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
There are no transcripts associated with this gene.

Gene
FERMT3
Gene Name
fermitin family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031471.5 Intron NP_113659.3
NM_178443.2 Intron NP_848537.1
XM_011545294.2 Intron XP_011543596.1
XM_011545295.2 Intron XP_011543597.1
XM_017018398.1 Intron XP_016873887.1
XM_017018399.1 Intron XP_016873888.1
Gene
NUDT22
Gene Name
nudix hydrolase 22
There are no transcripts associated with this gene.

Gene
TRPT1
Gene Name
tRNA phosphotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033678.3 Intron NP_001028850.2
NM_001160389.1 Intron NP_001153861.1
NM_001160390.1 Intron NP_001153862.1
NM_001160392.1 Intron NP_001153864.1
NM_001160393.1 Intron NP_001153865.1
NM_031472.3 Intron NP_113660.1
XM_005274345.4 Intron XP_005274402.1
XM_005274346.4 Intron XP_005274403.1
XM_005274347.4 Intron XP_005274404.1
XM_006718706.3 Intron XP_006718769.1
XM_006718707.3 Intron XP_006718770.1
XM_017018400.1 Intron XP_016873889.1
XM_017018401.1 Intron XP_016873890.1
Gene
VEGFB
Gene Name
vascular endothelial growth factor B
There are no transcripts associated with this gene.

View Full Product Details