Product Details

SNP ID
rs2999155
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:112679036 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATAAAAACCCTGGCTTTGACCC[G/A]CTAATGGGAGTTGGAATCCATAGTC
Phenotype
MIM: 601580 MIM: 610742
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CAPZA1 PubMed Links

Gene Details

Gene
CAPZA1
Gene Name
capping actin protein of muscle Z-line alpha subunit 1
There are no transcripts associated with this gene.

Gene
MOV10
Gene Name
Mov10 RISC complex RNA helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130079.2 Intron NP_001123551.1
NM_001286072.1 Intron NP_001273001.1
NM_001321324.1 Intron NP_001308253.1
NM_020963.4 Intron NP_066014.1
XM_005270869.4 Intron XP_005270926.1
XM_005270870.4 Intron XP_005270927.1
XM_017001318.1 Intron XP_016856807.1
XM_017001319.1 Intron XP_016856808.1

View Full Product Details