Product Details
- SNP ID
-
rs1127827
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:129095064 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCGCTGCAAGGTGCGCCTCGTGCCG[C/T]TGCAGATCCAGGTGGGTCCTGCGGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ISOC1
PubMed Links
Gene Details
- Gene
- ISOC1
- Gene Name
- isochorismatase domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016048.2 |
316 |
Silent Mutation |
CTG,TTG |
L100L |
NP_057132.2 |
- Gene
- MIR4633
- Gene Name
- microRNA 4633
There are no transcripts associated with this gene.
View Full Product Details