Product Details

SNP ID
rs2311426
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:26835073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCTTTAAGTTTGGGTGGAATTC[A/G]AGCACAGGAGCTGTGCGGAGGCAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZDHHC18 PubMed Links

Gene Details

Gene
ZDHHC18
Gene Name
zinc finger DHHC-type containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032283.2 Intron NP_115659.1

View Full Product Details