Product Details

SNP ID
rs2331821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69842743 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTACTGCCAGCCAGAGAGTGTTTCT[C/T]ATCTGGGTAATTATAATAGTCCAGC
Phenotype
MIM: 600043
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SULT1E1 PubMed Links

Gene Details

Gene
SULT1E1
Gene Name
sulfotransferase family 1E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005420.2 Intron NP_005411.1
XM_011532210.2 Intron XP_011530512.1

View Full Product Details