Product Details
- SNP ID
-
rs2344933
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:6497500 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCTGCTCCTCGGCTTTCTGTCTTC[A/T]CGACCTCCTTAGGCTGTGGGCGAAG
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DNHD1
PubMed Links
Gene Details
- Gene
- DNHD1
- Gene Name
- dynein heavy chain domain 1
View Full Product Details