Product Details

SNP ID
rs2345388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66726311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAACAAGCATTAAATAAAACTG[G/T]TTTTTAATGATTTTACTGCAGGCAG
Phenotype
MIM: 611406
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DYNC1LI2 PubMed Links

Gene Details

Gene
DYNC1LI2
Gene Name
dynein cytoplasmic 1 light intermediate chain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286157.1 Intron NP_001273086.1
NM_001323955.1 Intron NP_001310884.1
NM_006141.2 Intron NP_006132.1
XM_017023008.1 Intron XP_016878497.1

View Full Product Details