Product Details

SNP ID
rs2347279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:2538546 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGCAGACTGATGCCAGGAAAGAA[A/C]AAAGGCCTCTGTGTAGCCAGAGCCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
METTL4 PubMed Links

Gene Details

Gene
METTL4
Gene Name
methyltransferase like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308401.1 2288 UTR 3 NP_001295330.1
NM_022840.4 2288 UTR 3 NP_073751.3
XM_005258132.3 2288 UTR 3 XP_005258189.1
XM_005258133.2 2288 UTR 3 XP_005258190.1
XM_011525730.2 2288 UTR 3 XP_011524032.1

View Full Product Details