Product Details

SNP ID
rs3087565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:121180703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTTCTTCATTAAAATAATATA[A/C]ACAGAATGTATTGTTAGTTCGATTC
Phenotype
MIM: 605496 MIM: 612673
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CNTRL PubMed Links

Gene Details

Gene
CNTRL
Gene Name
centriolin
There are no transcripts associated with this gene.

Gene
RAB14
Gene Name
RAB14, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016322.3 1797 UTR 3 NP_057406.2

View Full Product Details