Product Details

SNP ID
rs2448243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:101967267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATTTGTACATTTAATCAAATAT[A/G]TGCATCCAGTAGTTAATTACAGTGA
Phenotype
MIM: 600112
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DYNC1H1 PubMed Links

Gene Details

Gene
DYNC1H1
Gene Name
dynein cytoplasmic 1 heavy chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001376.4 Intron NP_001367.2

View Full Product Details