Product Details
- SNP ID
-
rs2118139
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:66304658 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GATTAAAGATAGTGGTAAGTTTGAT[A/G]AAACTTTGAGGAAACGGTTTCTAAA
- Phenotype
-
MIM: 614539
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HELB
PubMed Links
Gene Details
- Gene
- HELB
- Gene Name
- DNA helicase B
View Full Product Details