Product Details

SNP ID
rs2846356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102691344 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACTTGTCAGAGAATAACCTCTAA[C/T]GTAACTCACCCTGCATCTATCCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP27 PubMed Links

Gene Details

Gene
MMP27
Gene Name
matrix metallopeptidase 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022122.2 1906 Intron NP_071405.2
XM_011542948.2 1906 UTR 3 XP_011541250.1
XM_011542949.2 1906 UTR 3 XP_011541251.1
XM_011542950.2 1906 UTR 3 XP_011541252.1
XM_017018120.1 1906 UTR 3 XP_016873609.1

View Full Product Details