Product Details

SNP ID
rs2846392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57377339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTGGCACCCATGCTGCTGCCCA[C/T]GTCTGCTGTTGGAGGTGGGCATCTT
Phenotype
MIM: 600843 MIM: 605601 MIM: 606814
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P2RX3 PubMed Links

Gene Details

Gene
P2RX3
Gene Name
purinergic receptor P2X 3
There are no transcripts associated with this gene.

Gene
PRG2
Gene Name
proteoglycan 2, pro eosinophil major basic protein
There are no transcripts associated with this gene.

Gene
PRG3
Gene Name
proteoglycan 3, pro eosinophil major basic protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006093.3 Intron NP_006084.2

View Full Product Details