Product Details

SNP ID
rs2852780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:44697023 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGTCCTGCAGGCAGTCATAGAAG[A/G]GTTAACTTTTGTTGGACTGAAGGCC
Phenotype
MIM: 611060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SETBP1 PubMed Links

Gene Details

Gene
SETBP1
Gene Name
SET binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130110.1 Intron NP_001123582.1
NM_015559.2 Intron NP_056374.2

View Full Product Details