Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001324171.1 | Intron | NP_001311100.1 | |||
NM_001324172.1 | Intron | NP_001311101.1 | |||
NM_030581.3 | Intron | NP_085058.3 | |||
XM_005256146.3 | Intron | XP_005256203.1 | |||
XM_011523331.2 | Intron | XP_011521633.1 | |||
XM_011523332.2 | Intron | XP_011521634.1 | |||
XM_017023667.1 | Intron | XP_016879156.1 | |||
XM_017023668.1 | Intron | XP_016879157.1 | |||
XM_017023669.1 | Intron | XP_016879158.1 | |||
XM_017023670.1 | Intron | XP_016879159.1 |