Product Details

SNP ID
rs2180950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:31519220 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAAATACATTCCTTTTAAATTGT[A/G]AATATATTTAGAGCATGTCCAGACT
Phenotype
MIM: 606985 MIM: 612323
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELP4 PubMed Links

Gene Details

Gene
ELP4
Gene Name
elongator acetyltransferase complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288725.1 Intron NP_001275654.1
NM_001288726.1 Intron NP_001275655.1
NM_019040.4 Intron NP_061913.3
Gene
IMMP1L
Gene Name
inner mitochondrial membrane peptidase subunit 1
There are no transcripts associated with this gene.

View Full Product Details