Product Details
- SNP ID
-
rs2168209
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:30630007 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTAATTTTTACATCTCTGTTTTTA[A/G]AAAAGATATATGTGAACTGGAAAAA
- Phenotype
-
MIM: 611299
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
G2E3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201656844] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- G2E3
- Gene Name
- G2/M-phase specific E3 ubiquitin protein ligase
There are no transcripts associated with this gene.
- Gene
- SCFD1
- Gene Name
- sec1 family domain containing 1
View Full Product Details