Product Details

SNP ID
rs2193931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151278219 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTTCTGCTGCTGTCCGAATATCA[A/G]CCACTTATTTTTCTATTCTTAATAG
Phenotype
MIM: 613109 MIM: 608332
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GM2A PubMed Links

Gene Details

Gene
GM2A
Gene Name
GM2 ganglioside activator
There are no transcripts associated with this gene.

Gene
SLC36A3
Gene Name
solute carrier family 36 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145017.1 Intron NP_001138489.1
NM_181774.3 Intron NP_861439.3
XM_011537627.2 Intron XP_011535929.1
XM_011537629.2 Intron XP_011535931.1
XM_011537630.1 Intron XP_011535932.1
XM_011537631.1 Intron XP_011535933.1
XM_011537633.1 Intron XP_011535935.1
XM_011537634.2 Intron XP_011535936.1

View Full Product Details