Product Details

SNP ID
rs2195153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:16588906 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATATACCAGGATGGCTAGTTTTAT[A/T]AAACAGTTTAGTTAAAAAAGAAATC
Phenotype
MIM: 601486
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DAZL PubMed Links

Gene Details

Gene
DAZL
Gene Name
deleted in azoospermia like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190811.1 Intron NP_001177740.1
NM_001351.3 Intron NP_001342.2

View Full Product Details