Product Details

SNP ID
rs2230566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3976551 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCGGCAGGCGCTGCAGGAAGGGC[C/T]GCCTACAATTTGTCCAGGAAGTTGT
Phenotype
MIM: 603289 MIM: 130610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAPK3 PubMed Links

Gene Details

Gene
DAPK3
Gene Name
death associated protein kinase 3
There are no transcripts associated with this gene.

Gene
EEF2
Gene Name
eukaryotic translation elongation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001961.3 2663 UTR 3 NP_001952.1
Gene
SNORD37
Gene Name
small nucleolar RNA, C/D box 37
There are no transcripts associated with this gene.

View Full Product Details