Product Details

SNP ID
rs2231579
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:86547103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAAACCAACATTTGAGCCAGGA[A/G]TAACTAGAGAGGAACAATGGGGTTA
Phenotype
MIM: 616857
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCA4 PubMed Links

Gene Details

Gene
CLCA4
Gene Name
chloride channel accessory 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012128.3 28 UTR 5 NP_036260.2
XM_011541015.2 28 Intron XP_011539317.1

View Full Product Details