Product Details
- SNP ID
-
rs2069951
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:35175961 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTCCCTTTTTTCATCACAGTCCCT[A/G]GCCCCTTCTTTCTTAGCCTCTAACA
- Phenotype
-
MIM: 600646
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PROCR
PubMed Links
Gene Details
- Gene
- PROCR
- Gene Name
- protein C receptor
View Full Product Details