Product Details

SNP ID
rs2073731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19450415 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGAGCTCCCCTCAAGCTGAAAG[C/T]GTGAAGAGGTGAGGAGGCAGCTATC
Phenotype
MIM: 601754
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C22orf39 PubMed Links

Gene Details

Gene
C22orf39
Gene Name
chromosome 22 open reading frame 39
There are no transcripts associated with this gene.

Gene
UFD1L
Gene Name
ubiquitin fusion degradation 1 like (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035247.2 Intron NP_001030324.2
NM_005659.6 Intron NP_005650.2
XM_011530361.2 Intron XP_011528663.1

View Full Product Details