Product Details
- SNP ID
-
rs2070799
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:547869 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCTCGGGAATCGACCTGGTTCCC[G/C]GGGATCAGGAAGCCGCTAAATGACC
- Phenotype
-
MIM: 116948
MIM: 600311
- Polymorphism
- G/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CDC34
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75799132] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDC34
- Gene Name
- cell division cycle 34
There are no transcripts associated with this gene.
- Gene
- GZMM
- Gene Name
- granzyme M
View Full Product Details