Product Details

SNP ID
rs2941736
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:63449986 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAAAATACTTCAGTGGGTGGTTA[A/G]AATGAAGAGGCAATATCAAACTATT
Phenotype
MIM: 605494
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGB3BP PubMed Links

Gene Details

Gene
ITGB3BP
Gene Name
integrin subunit beta 3 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206739.1 Intron NP_001193668.1
NM_014288.4 Intron NP_055103.3
XM_006710514.3 Intron XP_006710577.3
XM_006710515.3 Intron XP_006710578.3
XM_017000853.1 Intron XP_016856342.1
XM_017000854.1 Intron XP_016856343.1
XM_017000855.1 Intron XP_016856344.1
XM_017000856.1 Intron XP_016856345.1
XM_017000857.1 Intron XP_016856346.1

View Full Product Details