Product Details

SNP ID
rs2958659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40004908 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTACTGAGCTCCAGGTGAACAGC[A/G]GCTTCCTGGGAATTGATGGCTGAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM83F PubMed Links

Gene Details

Gene
FAM83F
Gene Name
family with sequence similarity 83 member F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138435.2 Intron NP_612444.2

View Full Product Details