Product Details

SNP ID
rs2268186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41075475 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATCTGGTAACTTTTTTGCTAAAT[A/G]CCTTACAAGCTTCCACCAGTCTGCA
Phenotype
MIM: 189903 MIM: 614393
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NFYA PubMed Links

Gene Details

Gene
NFYA
Gene Name
nuclear transcription factor Y subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002505.4 Intron NP_002496.1
NM_021705.3 Intron NP_068351.1
Gene
OARD1
Gene Name
O-acyl-ADP-ribose deacylase 1
There are no transcripts associated with this gene.

View Full Product Details