Product Details

SNP ID
rs2281836
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:49598999 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGGAAGTTTCAAAGCCAGCTTGAC[C/G]TGGTTGTGGCCGTTGGGCGAGATGA
Phenotype
MIM: 609193
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100288910 PubMed Links

Gene Details

Gene
LOC100288910
Gene Name
uncharacterized LOC100288910
There are no transcripts associated with this gene.

Gene
LRR1
Gene Name
leucine rich repeat protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152329.3 303 UTR 5 NP_689542.2
NM_203467.1 303 UTR 5 NP_982292.1

View Full Product Details