Product Details

SNP ID
rs2275707
Assay Type
Functionally tested
NCBI dbSNP Submissions
82
Location
Chr.1:219914705 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GTATATGAGTATTTGTGTCATACTT[A/C]AGGAAACTGAAATATTCCCTCACTT
Phenotype
MIM: 611146
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC30A10 PubMed Links

Gene Details

Gene
SLC30A10
Gene Name
solute carrier family 30 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018713.2 2749 UTR 3 NP_061183.2
XM_006711437.3 2749 UTR 3 XP_006711500.2
XM_017001684.1 2749 UTR 3 XP_016857173.1

View Full Product Details