Product Details

SNP ID
rs2276017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65998621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCAGGCTGCCTCCTCCTCTTCA[T/C]TCTCCTCCTCACTCTCATGATACTG
Phenotype
MIM: 603811
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
BANF1 PubMed Links

Gene Details

Gene
BANF1
Gene Name
barrier to autointegration factor 1
There are no transcripts associated with this gene.

Gene
EIF1AD
Gene Name
eukaryotic translation initiation factor 1A domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242481.1 620 Missense Mutation AAT,AGT N159S NP_001229410.1
NM_001242482.1 620 Missense Mutation AAT,AGT N159S NP_001229411.1
NM_001242483.1 620 Missense Mutation AAT,AGT N159S NP_001229412.1
NM_001242484.1 620 Missense Mutation AAT,AGT N159S NP_001229413.1
NM_001242485.1 620 Missense Mutation AAT,AGT N159S NP_001229414.1
NM_001242486.1 620 Missense Mutation AAT,AGT N159S NP_001229415.1
NM_032325.3 620 Missense Mutation AAT,AGT N159S NP_115701.2
XM_017018412.1 620 Missense Mutation AAT,AGT N159S XP_016873901.1

View Full Product Details