Product Details

SNP ID
rs2979870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:122068416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGGAGTTACCAAACTTATAGCA[A/C]AGGTCCTTCTGATCTCACCACATTT
Phenotype
MIM: 153455 MIM: 610479
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOX PubMed Links

Gene Details

Gene
LOX
Gene Name
lysyl oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178102.2 Intron NP_001171573.1
NM_001317073.1 Intron NP_001304002.1
NM_002317.6 Intron NP_002308.2
Gene
SRFBP1
Gene Name
serum response factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152546.2 Intron NP_689759.2
XM_017009111.1 Intron XP_016864600.1

View Full Product Details