Product Details

SNP ID
rs2289987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:36276641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGAATACAGGTGCTACCACTTA[C/T]CTAACCTCTGAGCTTCTGTTTGCTC
Phenotype
MIM: 612299
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COMMD9 PubMed Links

Gene Details

Gene
COMMD9
Gene Name
COMM domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101653.1 Intron NP_001095123.1
NM_001307932.1 Intron NP_001294861.1
NM_001307937.1 Intron NP_001294866.1
NM_014186.3 Intron NP_054905.2
XM_017017625.1 Intron XP_016873114.1

View Full Product Details