Product Details

SNP ID
rs2277103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28141855 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCCTAGAGAGCTTCCGGAAGTGC[C/T]TTTGGGGCAAGATGCGTGCTGGCCG
Phenotype
MIM: 602240
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZKSCAN8 PubMed Links

Gene Details

Gene
ZKSCAN8
Gene Name
zinc finger with KRAB and SCAN domains 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278119.1 38 Intron NP_001265048.1
NM_001278121.1 38 Intron NP_001265050.1
NM_001278122.1 38 Intron NP_001265051.1
NM_006298.3 38 Intron NP_006289.2
XM_011514870.2 38 UTR 5 XP_011513172.1
XM_011514873.2 38 UTR 5 XP_011513175.1
XM_011514874.2 38 Intron XP_011513176.1
XM_017011266.1 38 Intron XP_016866755.1
Gene
ZSCAN16-AS1
Gene Name
ZSCAN16 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details