Product Details

SNP ID
rs2290679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:111224715 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTGTCCCTCTCGCAGCGACGGCT[C/T]GGAGGGTGCGGGAGCCTGCCTTCGT
Phenotype
MIM: 114080
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAMK4 PubMed Links

Gene Details

Gene
CAMK4
Gene Name
calcium/calmodulin dependent protein kinase IV
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323374.1 Intron NP_001310303.1
NM_001323375.1 Intron NP_001310304.1
NM_001323376.1 Intron NP_001310305.1
NM_001323377.1 Intron NP_001310306.1
NM_001744.5 Intron NP_001735.1

View Full Product Details