Product Details

SNP ID
rs2481866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:39186694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGTTCACTTCAAGTTCTTCCTA[C/G]CCTGTACACTTCCCATTATATTCTT
Phenotype
MIM: 605296
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BRWD1 PubMed Links

Gene Details

Gene
BRWD1
Gene Name
bromodomain and WD repeat domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007246.2 7590 Intron NP_001007247.1
NM_018963.4 7590 UTR 3 NP_061836.2
NM_033656.3 7590 UTR 3 NP_387505.1
XM_011529611.1 7590 UTR 3 XP_011527913.1
XM_011529612.2 7590 UTR 3 XP_011527914.1
XM_011529613.1 7590 UTR 3 XP_011527915.1
XM_017028373.1 7590 UTR 3 XP_016883862.1
XM_017028374.1 7590 UTR 3 XP_016883863.1
XM_017028375.1 7590 Intron XP_016883864.1
Gene
PSMG1
Gene Name
proteasome assembly chaperone 1
There are no transcripts associated with this gene.

View Full Product Details