Product Details

SNP ID
rs2521795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19502819 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGCTTCCTAAAAAGCCTGGAAAA[A/G]TGCAACTCTGCCCCCTGGTGGTTTG
Phenotype
MIM: 605943
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GDE1 PubMed Links

Gene Details

Gene
GDE1
Gene Name
glycerophosphodiester phosphodiesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324066.1 1811 UTR 3 NP_001310995.1
NM_001324067.1 1811 UTR 3 NP_001310996.1
NM_016641.3 1811 UTR 3 NP_057725.1
Gene
TMC5
Gene Name
transmembrane channel like 5
There are no transcripts associated with this gene.

View Full Product Details